Order Entry
Switzerland
ContactUsLinkComponent
 
Human recombinant LAMP2 (from cells)
Catalog # PRSI91-437
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant LAMP2 (from cells)
Catalog # PRSI91-437
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    LAMP-2|Lysosome-Associated Membrane Glycoprotein 2|CD107 Antigen-like Family Member B|Lysosome-Associated Membrane Protein 2|LAMP2|CD107b
  • Protein/peptide name:
    LAMP2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    33.94 kD
  • Sequence:
    Leu29-Ile375
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Lysosomal Associated Membrane Protein 2 (LAMP2) is a major component of lysosomal membranes. LAMP2 is a transmembrane glycoprotein about 110kDa. Mature human LAMP2 consists of a 347 amino acid (aa) intralumenal domain, a 24 aa transmembrane segment, and a 35 aa cytoplasmic tail . The lumenal domain is organized into two heavily N-glycosylated regions. Alternate splicing generates a human LAMP2 isoform (LAMP2B) with a substituted juxtamembrane lumenal region, cytoplasmic tail and transmenmbrane segment.LAMP2 itself can cleavage lysosomal luminal domain and degradation lysosomal. In the help of chaperone HSC73,LAMP2 mediates the lysosomal uptake in complex with cargo proteins and is required for the lysosomal destruction of autophagic vacuoles.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag