- Pk:50 µG
- Protein/peptide type:Recombinant
- Source:Cells
- Species:Human
- Tag sequence:C-6 His tag
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Protein synonyms:IL-1 Receptor Accessory Protein|IL1RAP|Interleukin-1 Receptor Accessory Protein|IL-1R3|IL-1R-3|IL1R3|IL-1RAcP|Interleukin-1 Receptor 3|C3orf13
- Protein/peptide name:IL-1RAcP
- Purity:> 95% as determined by reducing SDS-PAGE
- Molecular weight:39.75 kD
- Sequence:Ser21-Gln356
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tested applications:Bioassay
Interleukin-1 Receptor Accessory Protein (IL-1RAcP) is a member of the interleukin-1 receptor family. It contains three Ig-like C2-type domains in the extracellular region and a long cytoplasmic domain implicated in signal transduction. IL-1RAcP acts as a non-ligand binding accessory component of the receptors for IL1 alpha, IL1 beta and IL33. IL-1RAcP mediates interleukin-1-dependent activation of NF-kappa-B. It is part of the membrane-bound form of the IL-1 receptor. IL-1 RAcP takes part in the Signaling ways by the formation of a ternary complex containing IL1R1, TOLLIP, MYD88, and IRAK1 or IRAK2. In addition, IL-1RAcP modulates the response to interleukins by associating with soluble IL1R1 and enhancing interleukin-binding to the decoy receptor.
- This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag