Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant Ephrin B2 (from cells)
Human recombinant Ephrin B2 (from cells)
Catalog # PRSI91-421
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Ephrin B2 (from cells)
Catalog # PRSI91-421
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    EPH-Related Receptor Tyrosine Kinase Ligand 5|EFNB2|EPLG5|HTK-L|HTKL|LERK-5|LERK5|HTK Ligand|Ephrin-B2
  • Protein/peptide name:
    Ephrin B2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    23.24 kD
  • Sequence:
    Ile28-Ala229
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    For Biological Assays

Specifications

About this item

Ephrin-B2 is a type I transmembrane protein and belongs the Ephrin family. It binds to the receptor tyrosine kinases, such as EPHA4, EPHB4 and EPHA3. Ephrin-B2 has been implicated in mediating developmental events, especially in the nervous system, erythropoiesis and tumour metastasis. Ligation of Ephrin-B2 with complementary EphB receptors on adjacent cells results in a combination of forward (EphB receptors) and reverse (Ephrin-B2) signalling, which is central to tissue development and remodelling functions. In addition, Ephrin-B2 may play a role in constraining the orientation of longitudinally projecting axons.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag