Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant Clusterin (from cells)
Human recombinant Clusterin (from cells)
Catalog # PRSI91-410
Supplier:  ProSci Inc.
CAS Number:  
Human recombinant Clusterin (from cells)
Catalog # PRSI91-410
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    Cells
  • Species:
    Human
  • Tag sequence:
    C-6 His tag
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Protein synonyms:
    Apolipoprotein J|NA1/NA2|KUB1|TRPM-2|Clusterin|Complement-Associated Protein SP-40|APOJ|Apo-J|CLI|Aging-Associated Gene 4 Protein|CLU|Complement Cytolysis Inhibitor|40|Ku70-Binding Protein 1|Testosterone-Repressed Prostate Message 2
  • Protein/peptide name:
    Clusterin
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Molecular weight:
    51.1 kD
  • Sequence:
    Asp23-Glu449
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tested applications:
    Bioassay

Specifications

About this item

Clusterin is a secreted protein which belongs to the Clusterin family. Clusterin is expressed in adult testis, heart, ovary, adrenal gland, brain and liver. Clusterin has been suggested to be involved in several basic biological events such as cell death, tumor progression, and neurodegenerative disorders. In addition,Clusterin is up/ down regulated on the mRNA or protein level in many pathological and clinically relevant situations including cancer, organ regeneration, infection, Alzheimer disease, retinitis pigmentosa, myocardial infarction, renal tubular damage, autoimmunity and others.

  • This recombinant protein can be used for biological assays. For research use only.

Fusion-Tag: C-6 His tag