Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant VSIG2 (from HEK293 cells)
Human recombinant VSIG2 (from HEK293 cells)
Catalog # PRSI96-779
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant VSIG2 (from HEK293 cells)
Catalog # PRSI96-779
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    200 µG
  • Protein/peptide type:
    Recombinant
  • Source:
    HEK293 cells
  • Species:
    Human
  • Protein synonyms:
    CTXL|CTH|VSIG2|CT-like protein
  • Protein/peptide name:
    VSIG2
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular weight:
    24 kDa
  • Formulation:
    Lyophilized, PBS, pH7.4
  • Tested applications:
    Western Blot

Specifications

About this item

V-set and immunoglobulin domain-containing protein 2 (VSIG2) is also known as Cortical thymocyte-like protein, CTH, CTXL, is a single-pass type I membrane protein which contains Ig-like C2-type (immunoglobulin-like) domain and one Ig-like V-type (immunoglobulin-like) domain. V-set domains are Ig-like domains resembling the antibody variable domain. VSIG2 is highly expressed in stomach, colon, prostate, trachea and thyroid glands and weakly in bladder and lung. V-set domains are found in diverse protein families, including immunoglobulin light and heavy chains.