Auftragserfassung
Switzerland
ContactUsLinkComponent
Human recombinant VLDLR (from HEK293 cells)
Human recombinant VLDLR (from HEK293 cells)
Catalog # PRSI96-778
Supplier:  ProSci Inc.
CAS Number:  
undefined
Human recombinant VLDLR (from HEK293 cells)
Catalog # PRSI96-778
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • VE:
    100 µG
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    HEK293 cells
  • Spezies:
    Human
  • Biologische Aktivität:
    Measured by its binding ability in a functional ELISA. When Recombinant Human Apolipoprotein E3 is immobilized at 1μg/ml (100μl/well), the concentration of Recombinant Human VLDLR that produces 50% of the optimal binding response is found to be approximately 0. 03 - 0. 15μg/ml.
  • Proteinsynonym:
    RP11-320E16.1|VLDL receptor|VLDLRCH|FLJ35024|VLDLR|CHRMQ1
  • Protein/Peptid-Name:
    VLDLR
  • Purity:
    >97% as determined by SDS-PAGE.
  • Molecular weight:
    86 kDa
  • Zusammensetzung:
    Lyophilized, PBS, pH7.4
  • Validierte Anwendungen:
    Western Blot

Specifications

About this item

The very-low-density-lipoprotein receptor (VLDL-R) is a lipoprotein receptor that shows considerable similarity to the lowdensity-lipoprotein receptor. VLDL R is a 130 kDa type I transmembrane protein in the LDL receptor family that plays a significant role in lipid metabolism and in nervous system development and function .This receptor has been suggested to be important for the metabolism of apoprotein-E-containing triacylglycerol-rich lipoproteins, such as very-low-densitylipoprotein (VLDL), beta-migrating VLDL and intermediate-density lipoprotein. It is also one of the receptors of reelin, an extracellular matrix protein which regulates the processes of neuronal migration and synaptic plasticity. In humans, the VLDL-R is encoded by the VLDLR gene.
A rare neurological disorder first described in the 1970s under the name "disequilibrium syndrome" is now considered to be caused by the disruption of VLDLR gene. The disorder was renamed VLDLR-associated cerebellar hypoplasia (VLDLRCH) after a 2005 study. It is associated with parental consanguinity and found in secluded communities such as the Hutterites. VLDLRCH is one of the two known genetic disorders caused by a disruption of reelin signaling pathway, along with Norman-Roberts syndrome.