Human recombinant G-CSF (from E. coli)
Catalog # PRSI40-555
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:10 µG
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Biological activity:Biological activity was determined by its ability to stimulate the proliferation of mouse M-NFS-60 cells. The expected ED50 is ≤ 0.1 ng/mL, corresponding to a specific activity of ≥ 1 x 107 units/mg. Endotoxin level is less than 0.1 ng per μg (1EU/μg).
- Gene ID:1440
- Protein synonyms:G-CSF|SCN1|PMN-E|C17orf33|HLE|HNE|GCSF|NE|ELA2|Granulocyte colony-stimulating factor|GE|Pluripoietin
- Protein/peptide name:G-CSF
- Purity:>98%
- Formulation:Lyophilized
Specifications
About this item
G-CSF is a hematopoietic growth factor that stimulates the development of committed progenitor cells to neutrophils and enhances the functional activities of the mature end-cell. It is produced in response to specific stimulation by a variety of cells including macrophages, fibroblasts, endothelial cells and bone marrow stroma. G-CSF is being used clinically to facilitate hematopoietic recovery after bone marrow transplantation. Human and murine G-CSF are cross-species reactive. Recombinant human G-CSF is an 18.7 kDa protein consisting of 174 amino acid residues.
- Store in working aliquots at -20°C. Avoid freeze/thaw cycles.