Saccharomyces cerevisiae S288c A synthetic peptide from the amino terminus of yeast Pst1
Catalog # PRSI3825P
Supplier: ProSci Inc.
CAS Number:
Specifications
- Pk:50 µG
- Species:Saccharomyces cerevisiae S288c
- Storage conditions:Store at –20 °C, stable for one year.
- Protein/peptide name:PST1 Peptide (amino terminus)
- Tested applications:Blocking
Specifications
About this item
Pst1 peptide is used for blocking the activity of Pst1 antibody.