Order Entry
Switzerland
ContactUsLinkComponent
SARS M protein SARS M protein peptide (13 aa near C-terminus)
SARS M protein SARS M protein peptide (13 aa near C-terminus)
Catalog # PRSI3529P
Supplier:  ProSci Inc.
CAS Number:  
SARS M protein SARS M protein peptide (13 aa near C-terminus)
Catalog # PRSI3529P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    SARS M protein
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    13 aa near C-terminus
  • Protein/peptide name:
    SARS M protein Peptide (13 aa near C-terminus)
  • Tested applications:
    Blocking

Specifications

About this item

SARS matrix peptide is used for blocking the activity of the SARS matrix antibody.