Order Entry
Switzerland
ContactUsLinkComponent
 
Human ARMER peptide (15 aa near N-terminus)
Catalog # PRSI3305P
Supplier:  ProSci Inc.
CAS Number:  
Human ARMER peptide (15 aa near N-terminus)
Catalog # PRSI3305P
Supplier:  ProSci Inc.
CAS Number:  

Specifications

  • Pk:
    50 µG
  • Species:
    Human
  • Storage conditions:
    Store at –20 °C, stable for one year.
  • Amino acid number:
    15 aa near N-terminus
  • Protein/peptide name:
    ARMER Peptide (15 aa near N-terminus)
  • Tested applications:
    Blocking

Specifications

About this item

ARMER peptide is used for blocking the activity of ARMER antibody.