Anti-PDGFB Rabbit Polyclonal Antibody (Biotin)
Catalog # PRSIXP-5255BT
Supplier: ProSci Inc.
Specifications
- Antigen name:Platelet-derived Growth Factor Beta
- Antigen symbol:PDGFB
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:SIS|PDGF subunit B|PDGF2|c-sis|SSV|PDGF-B|Platelet-derived growth factor subunit B|PDGF-2|IBGC5
- Immunogen:Produced from sera of rabbits pre-immunized with highly pure (>98%) recombinant hPDGF-BB (human Platelet Derived Growth Factor-BB).
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Anti-hPDGF-BB specific antibody was purified by affinity chromatography and then biotinylated.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PDGFB
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human