Order Entry
Switzerland
ContactUsLinkComponent
Anti-DNM1 Rabbit Polyclonal Antibody
Anti-DNM1 Rabbit Polyclonal Antibody
Catalog # PRSI79-752
Supplier:  ProSci Inc.
undefined
Anti-DNM1 Rabbit Polyclonal Antibody
Catalog # PRSI79-752
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Dynamin 1
  • Antigen symbol:
    DNM1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    EC 3.6.5.5|DNM1|Dynamin BREDNM19|DNM|brain|dynamin-1|Dynamin|B-dynamin|D100
  • Concentration:
    1 mg/mL
  • Immunogen:
    Dynamin 1 antibody  was raised against a peptide sequence around aa.774~778 (S-P-T-S-S) derived from Human Dynamin 1 .
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Microtubule-associated force-producing protein involved in producing microtubule bundles and able to bind and hydrolyze GTP. Most probably involved in vesicular trafficking processes. Involved in receptor-mediated endocytosis

Western Blot: 1:500~1:1000

Antibodies were purified by affinity-chromatography using epitope-specific peptide.

Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: DNM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat