Order Entry
Switzerland
ContactUsLinkComponent
Anti-JUP Rabbit Polyclonal Antibody
Anti-JUP Rabbit Polyclonal Antibody
Catalog # PRSI79-746
Supplier:  ProSci Inc.
undefined
Anti-JUP Rabbit Polyclonal Antibody
Catalog # PRSI79-746
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Plakoglobin
  • Antigen symbol:
    JUP
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Form:
    Liquid
  • Antigen synonyms:
    PKGB|DPIII|gamma (80kD)|desmoplakin-3|PDGB|catenin gamma|desmoplakin III|DP3|gamma 80kDa|CTNNG|catenin (Cadherin-associated protein)|ARVD12|gamma-catenin
  • Concentration:
    1 mg/mL
  • Immunogen:
    gamma-Catenin antibody was raised in was raised against a peptide sequence around aa.726~730 (I-D-T-Y-S) derived from Human γ-Catenin.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Common junctional plaque protein. The membrane-associated plaques are architectural elements in an important strategic position to influence the arrangement and function of both the cytoskeleton and the cells within the tissue. The presence of plakoglobin in both the desmosomes and in the intermediate junctions suggests that it plays a central role in the structure and function of submembranous plaques. Acts as a substrate for VE-PTP and is required by it to stimulate VE-cadherin function in endothelial cells. Can replace beta-catenin in E-cadherin/catenin adhesion complexes which are proposed to couple cadherins to the actin cytoskeleton

Western Blot: 1:500~1:1000

Antibodies were purified by affinity-chromatography using epitope-specific peptide.

Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: JUP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Rat