Anti-PRDM1 Rabbit Polyclonal Antibody
Catalog # PRSI79-744
Anbieter: ProSci Inc.
Technische Daten
- Antigen name:PR Domain zinc Finger protein 1
- Antigen symbol:PRDM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:PRDI-BF1|BLIMP1
- Concentration:1 mg/mL
- Immunogen:Blimp-1 antibody was raised against a peptide sequence around aa.124~128 (D-T-V-P-K) derived from Human Blimp-1.
- Size:100 µg
- Pk:100 µG
Technische Daten
Über diesen Artikel
Transcriptional repressor that binds specifically to the PRDI element in the promoter of the beta-interferon gene. Drives the maturation of B-lymphocytes into Ig secreting cells.
Western Blot: 1:500~1:1000
Antibodies were purified by affinity-chromatography using epitope-specific peptide.
Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: PRDM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat