Anti-FFAR1 Rabbit Polyclonal Antibody
Catalog # PRSI71-310
Supplier: ProSci Inc.
Specifications
- Antigen name:Free Fatty Acid Receptor 1
- Antigen symbol:FFAR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Concentration:1 mg/mL
- Immunogen:Synthetic peptide containing a sequence corresponding to a region within amino acids 239 and 300 of Human GPR40.
- Tested applications:Immunoaffinity Chromatography
- Size:50 µl
- Pk:50 µl
Specifications
About this item
FFAR1 / GPR40 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 10 μg/mL, and Western Blotting starting at 1:500 - 1:3000.
PBS, pH 7, 1% BSA, 20% Glycerol, 0.01% Thimerosal
PBS, pH 7, 1% BSA, 20% Glycerol, 0.01% Thimerosal
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: FFAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human