Anti-C9 Rabbit Polyclonal Antibody
Catalog # PRSI70-848
Supplier: ProSci Inc.
Specifications
- Antigen symbol:C9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human
- Western blot:Yes
- Concentration:1 mg/mL
- Immunogen:Recombinant protein fragment containing a sequence corresponding to a region within amino acids 184 and 411 of Complement C9.
- Tested applications:Immunoaffinity Chromatography
- Size:50 µl
- Pk:50 µl
Specifications
About this item
C9 Antibody can be used for Immunocytochemistry starting at 1:100 - 1:200, Immunofluorescence starting at 1:100 - 1:200, Immunohistochemistry in paraffin embeded tissues starting at 10μg/mL, and Western Blotting starting at 1:500 - 1:3000.
0.1 M Tris-glycine, pH 7, 10% Glycerol, 0.01% Thimerosal
0.1 M Tris-glycine, pH 7, 10% Glycerol, 0.01% Thimerosal
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type: 0
Antigen: C9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human