Anti-COPS5 Rabbit Polyclonal Antibody
PRSI70-800
- Antigen name:COP9 Signalosome Subunit 5
- Antigen symbol:COPS5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:JAB1|CSN5|SGN5|MOV-34
- Concentration:0.2 mg/mL
- Immunogen:A synthetic peptide from N-Terminal of human JAB1 protein.
- Tested applications:Immunoaffinity Chromatography
- Size:50 µg
- Pk:250 µl
COPS5 Antibody can be used for Immunohistochemistry in paraffin embeded tissues starting at 1:100, and Western Blotting starting at 1 - 2 μg/mL.
10 mM PBS, pH 7.4, BSA, sodium azide.
10 mM PBS, pH 7.4, BSA, sodium azide.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: COPS5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: