Anti-UCHL1 Rabbit Polyclonal Antibody
PRSI70-660
- Antigen name:Ubiquitin Carboxyl-Terminal Esterase L1
- Antigen symbol:UCHL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:Neuron cytoplasmic protein 9.5|Ubiquitin thioesterase L1.|PGP9.5|Ubiquitin carboxyl-terminal|hydrolase isozyme L1|Ubiquitin thioesterase L1|UCH-L1|PGP 9.5|PARK5
- Concentration:1 mg/mL
- Immunogen:Synthetic peptide
- Tested applications:Immunoaffinity Chromatography
- Size:50 µg
- Pk:50 µl
MMP9 Antibody can be used for ELISA starting at 1:1000, and Immunohistochemistry in paraffin embeded tissues starting at 10 μg/mL,
PBS (without Mg2+, Ca2+), pH 7.4, 150 mM NaCl, 0.02% sodium azide, 50% glycerol
PBS (without Mg2+, Ca2+), pH 7.4, 150 mM NaCl, 0.02% sodium azide, 50% glycerol
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat