Anti-FFAR2 Rabbit Polyclonal Antibody
Catalog # PRSI70-655
Supplier: ProSci Inc.
Specifications
- Antigen name:Free Fatty Acid Receptor 2
- Antigen symbol:FFAR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Antigen synonyms:FFA2R|GPR43
- Concentration:1 mg/mL
- Immunogen:Synthetic peptide
- Tested applications:Immunoaffinity Chromatography
- Size:50 µg
- Pk:50 µl
Specifications
About this item
FFAR2 / GPR43 Antibody can be used for ELISA starting at 1:1000, and Immunofluorescence starting at 1:100 - 1:500,
PBS (without Mg2+, Ca2+), pH 7.4, 150 mM NaCl, 0.02% sodium azide, 50% glycerol
PBS (without Mg2+, Ca2+), pH 7.4, 150 mM NaCl, 0.02% sodium azide, 50% glycerol
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: FFAR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human