Anti-CTSL2 Rabbit Polyclonal Antibody
Catalog # PRSI70-642
Supplier: ProSci Inc.
Specifications
- Antigen symbol:CTSL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Concentration:1 mg/mL
- Immunogen:Synthetic peptide from human CATL2.
- Tested applications:Immunoaffinity Chromatography
- Size:50 µg
- Pk:50 µl
Specifications
About this item
ACVR2A Antibody can be used for ELISA starting at 1:1000, and Immunohistochemistry in paraffin embeded tissues starting at 5 μg/mL,
PBS (without Mg2+, Ca2+), pH 7.4, 150 mM NaCl, 0.02% sodium azide, 50% glycerol
PBS (without Mg2+, Ca2+), pH 7.4, 150 mM NaCl, 0.02% sodium azide, 50% glycerol
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: CTSL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: