Anti-SRSF3 Rabbit Polyclonal Antibody
PRSI70-641
- Antigen name:Serine/Arginine-Rich Splicing Factor 3
- Antigen symbol:SRSF3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Antigen synonyms:SFRS3|Serine/arginine-rich splicing factor 3|Pre-mRNA-splicing factor SRP20|Splicing factor arginine/serine-rich 3|SRP20
- Concentration:1 mg/mL
- Immunogen:Synthetic peptide from human SFRS3.
- Tested applications:Immunoaffinity Chromatography
- Size:50 µg
- Pk:50 µl
SRSF3 / SFRS3 Antibody can be used for ELISA starting at 1:5000, Immunofluorescence starting at 1:100 - 1:500, Immunohistochemistry in paraffin embeded tissues starting at 5μg/mL, and Western Blotting starting at 1:500 - 1:1000.
PBS (without Mg2+, Ca2+), pH 7.4, 150 mM NaCl, 0.02% sodium azide, 50% glycerol
PBS (without Mg2+, Ca2+), pH 7.4, 150 mM NaCl, 0.02% sodium azide, 50% glycerol
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SRSF3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse