Anti-HAMP Mouse Monoclonal Antibody
PRSI51-590
- Antibody type:Tag
- Antigen symbol:HAMP
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Immunogen:HAMP (AAH20612, 25 a.a. ~ 85 a.a) full length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:50 µg
- Pk:50 µG
Hepcidin antibody can be used in Western Blot, immunohistochemistry starting at 1:50, and immunoprecipitation.
Protein A Column
PBS, pH 7.2
Store Hepcidin antibody at 4 °C or -20 °C. As with all antibodies avoid freeze/thaw cycles.
Type:
Antigen: HAMP
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1,k
Reactivity: Human