Anti-RUNX2 Mouse Monoclonal Antibody
PRSI51-436
- Antibody type:Tag
- Antigen name:Runt-related Transcription Factor 2
- Antigen symbol:RUNX2
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Rat
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:Acute myeloid leukemia 3 protein|PEBP2A|CBFA1|PEBP2-alpha A|SL3/AKV core-binding factor alpha A subunit|Polyomavirus enhancer-binding protein 2 alpha A subunit|OSF2|OSF-2|Core-binding factor subunit alpha-1|RUNX2|Runt-related transcription factor 2|PEA2-alpha A|CBF-alpha-1|Osteoblast-specific transcription factor 2|SL3-3 enhancer factor 1 alpha A subunit|AML3|Oncogene AML-3
- Immunogen:RUNX2 (NP_004339, 251 a.a. ~ 351 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Size:50 µg
- Pk:50 µG
RUNX2 antibody can be used in immunohistochemistry starting at 15 μg/mL.
Protein A Column
PBS, pH 7.2
Store RUNX2 antibody at -20 °C. Aliquot to avoid freeze/thaw cycles.
Type: Tag
Antigen: RUNX2
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human, Rat