Order Entry
Switzerland
ContactUsLinkComponent
Anti-ABCB1 Rabbit Polyclonal Antibody
Anti-ABCB1 Rabbit Polyclonal Antibody
Catalog # PRSI48-396
Supplier:  ProSci Inc.
undefined
Anti-ABCB1 Rabbit Polyclonal Antibody
Catalog # PRSI48-396
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    ATP-binding cassette, Subfamily B (MDR/TAP), Member 1
  • Antigen symbol:
    ABCB1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Gibbon,
    Gorilla,
    Human
  • Form:
    Liquid
  • Antigen synonyms:
    CD243|ABC20|MDR1|P-GP|PGY1|GP170|CLCS
  • Immunogen:
    ABCB1 antibody was raised against a peptide located in the cytoplasmic domain of ABCB1 (Human).
  • Tested applications:
    Immunoaffinity Chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Abcb1/MDR1/P glycoprotein is an ATP dependent transporter that mediates efflux of chemotherapeutic agents from the intracellular milieu. It has also been associated with volume-regulated chloride channel activity and basal to apical transendothelial migration of mononuclear phagocytes.

PBS, 0.1% sodium azide.

Aliquot and store at -80°C. Avoid multiple freeze-thaw cycles.

Type:
Antigen: ABCB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Gibbon, Gorilla, Human