Anti-BDH2 Goat Polyclonal Antibody
PRSI46-632
- Antigen name:3-Hydroxybutyrate Dehydrogenase, Type 2
- Antigen symbol:BDH2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:UNQ6308|UCPA-OR|3-hydroxybutyrate dehydrogenase|EFA6R|oxidoreductase UCPA|BDH2|DHRS6|Dehydrogenase/reductase (SDR family) Member 6|PRO20933|FLJ13261|type 2
- Concentration:500 ug/mL
- Immunogen:BDH2 antibody was raised against a 12 amino acid synthetic peptide near the internal region of BDH2.
- Size:100 µg
- Pk:100 µG
ELISA: Antibody detection limit dilution 1:64,000. Western Blot: Approximately 26 kDa band observed in human cerebellum, kidney and liver lysates (calculated MW of 26.7 kDa according to NP_064524.3). Recommended concentration: 0.3-1 μg/mL.
BDH2 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
BDH2 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: BDH2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human