Anti-Importin 8 Goat Polyclonal Antibody
Catalog # PRSI45-771
Supplier: ProSci Inc.
Specifications
- Antigen name:Importin 8
- Antigen symbol:IMP8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Form:Liquid
- Antigen synonyms:RANBP8
- Concentration:500 ug/mL
- Immunogen:Importin 8 antibody was raised against a 12 amino acid synthetic peptide near the C-Terminus of Importin 8.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Importin 8 antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
Importin 8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: Importin 8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human