Anti-ABCC8 Goat Polyclonal Antibody
Catalog # PRSI42-749
Supplier: ProSci Inc.
Specifications
- Antigen name:ATP-binding cassette, Subfamily C (CFTR/MRP), Member 8
- Antigen symbol:ABCC8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Antigen synonyms:SUR1|SUR|PHHI |MRP8|HRINS|sulfonylurea receptor 1|HI|HHF1|ABC36|TNDM2
- Concentration:500 ug/mL
- Immunogen:ABCC8 antibody was raised against a 13 amino acid synthetic peptide near the C-Terminus of ABCC8.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Antibody was purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
ABCC8 antibody is supplied in Tris saline, 0.02% sodium azide, pH 7.3 with 0.5% BSA.
Aliquot and store at -20°C. Avoid multiple freeze-thaw cycles.
Type:
Antigen: ABCC8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human