Order Entry
Switzerland
ContactUsLinkComponent
Anti-FOXK2 Rabbit Polyclonal Antibody
Anti-FOXK2 Rabbit Polyclonal Antibody
Catalog # PRSI31-177
Supplier:  ProSci Inc.
undefined
Anti-FOXK2 Rabbit Polyclonal Antibody
Catalog # PRSI31-177
Supplier:  ProSci Inc.

Specifications

  • Antigen symbol:
    FOXK2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    cellular transcription factor ILF-1|ILF|FOXK2|interleukin enhancer binding factor 1|ILF-1|ILF1|forkhead box K2
  • Immunogen:
    A synthetic peptide corresponding a region of human FOXK2.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

FOXK2 contains a fork head DNA binding domain. This protein can bind to the purine-rich motifs of the HIV long terminal repeat (LTR), and to the similar purine-rich motif in the interleukin 2 (IL2) promoter. It may be involved in the regulation of viral and cellular promoter elements. Three alternatively spliced transcript variants encoding distinct isoforms have been described.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: FOXK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human