Specifications
- Antigen-Name:Adaptor-related Protein Complex 2, Beta 1 Subunit
- Antigen-Symbol:AP2B1
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Rabbit
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen-Synonyme:AP2-BETA|AP105B|ADTB2|CLAPB1
- Immunogen:A synthetic peptide corresponding a region of human AP2B1.
- Größe:50 µg
- VE:50 µG
Specifications
About this item
AP2B1is one of two large chain components of the assembly protein complex 2, which serves to link clathrin to receptors in coated vesicles. AP2B1 is found on the cytoplasmic face of coated vesicles in the plasma membrane.The protein encoded by this gene is one of two large chain components of the assembly protein complex 2, which serves to link clathrin to receptors in coated vesicles. The encoded protein is found on the cytoplasmic face of coated vesicles in the plasma membrane. Two transcript variants encoding different isoforms have been found for this gene.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: AP2B1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat