Order Entry
Switzerland
ContactUsLinkComponent
Anti-RTN2 Rabbit Polyclonal Antibody
Anti-RTN2 Rabbit Polyclonal Antibody
Catalog # PRSI30-211
Supplier:  ProSci Inc.
undefined
Anti-RTN2 Rabbit Polyclonal Antibody
Catalog # PRSI30-211
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Reticulon 2
  • Antigen symbol:
    RTN2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    NSP2|NSPL1|NSPLI|SPG12
  • Immunogen:
    A synthetic peptide corresponding a region of human RTN2.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

This gene belongs to the family of reticulon encoding genes. Reticulons are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells.This gene belongs to the family of reticulon encoding genes. Reticulons are associated with the endoplasmic reticulum, and are involved in neuroendocrine secretion or in membrane trafficking in neuroendocrine cells. Alternatively spliced transcript variants encoding different isoforms have been identified.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: RTN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat