Specifications
- Antigen name:Apolipoprotein B mRNA Editing Enzyme, Catalytic Polypeptide-Like 3B
- Antigen symbol:APOBEC3B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:ARCD3|PHRBNL|APOBEC1L|A3B|DJ742C19.2|ARP4|bK150C2.2
- Immunogen:A synthetic peptide corresponding a region of human APOBEC3B.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
APOBEC3B is a member of the cytidine deaminase family. It is thought that these proteins may be RNA editing enzymes and have roles in growth or cell cycle control.This gene is a member of the cytidine deaminase gene family. It is one of seven related genes or pseudogenes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: APOBEC3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human