Anti-NR3C2 Rabbit Polyclonal Antibody
Catalog # PRSI30-075
Supplier: ProSci Inc.
Specifications
- Antigen name:Nuclear Receptor Subfamily 3 Group C Member 2
- Antigen symbol:NR3C2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:NR3C2VIT|MLR|MCR|MR
- Immunogen:A synthetic peptide corresponding a region of human NR3C2.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
NR3C2 is a receptor for both mineralocorticoids (MC) such as aldosterone and glucocorticoids (GC) such as corticosterone or cortisol. It binds to mineralocorticoid response elements (MRE) and transactivates target genes. The effect of MC is to increase ion and water transport and thus raise extracellular fluid volume and blood pressure and lower potassium levels.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: NR3C2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human