Anti-RSAD2 Rabbit Polyclonal Antibody
Catalog # PRSI29-792
Supplier: ProSci Inc.
Specifications
- Antigen name:Radical S-adenosyl Methionine Domain Containing 2
- Antigen symbol:RSAD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Immunogen:A synthetic peptide corresponding a region of human RSAD2.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
RSAD2 is a potential antiviral effector. Western blots using two different antibodies against two unique regions of this protein target confirm the same apparent molecular weight in our tests.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: RSAD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat