Anti-EXOSC7 Rabbit Polyclonal Antibody
Catalog # PRSI29-421
Supplier: ProSci Inc.
Specifications
- Antigen name:Exosome Component 7
- Antigen symbol:EXOSC7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:Rrp42p|p8|RRP42|EAP1|hRrp42p
- Immunogen:A synthetic peptide corresponding a region of human EXOSC7.
- Size:100 µg
- Pk:100 µG
Specifications
About this item
EXOSC7 belongs to the RNase PH family. It is a component of the exosome 3'->5' exoribonuclease complex and is required for the 3' processing of the 7S pre-RNA to the mature 5.8S rRNA.
Antibody is purified by protein A chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: EXOSC7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse, Rat