Anti-SCNN1B Rabbit Polyclonal Antibody
Catalog # PRSI28-219
Supplier: ProSci Inc.
Specifications
- Antigen name:Sodium Channel Nonvoltage-gated 1 Beta
- Antigen symbol:SCNN1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human,Dog
- Western blot:Yes
- Form:Lyophilized
- Antigen synonyms:ENaCb|BESC1|ENaCbeta|SCNEB
- Immunogen:A synthetic peptide corresponding a region of human SCNN1B.
- Size:50 µg
- Pk:50 µG
Specifications
About this item
When expressed in the bladder epithelium, the protein encoded by the SCNN1B gene might be implicated in the mechanosensory transduction in the bladder afferent pathways, thereby inducing detrusor instability by outlet obstruction. Common variants of this gene confer susceptibility to human essential hypertension.
Antibody is purified by peptide affinity chromatography method.
Antibody is lyophilized in PBS buffer with 2% sucrose. Add 50 μL of distilled water. Final antibody concentration is 1 mg/mL.
Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
Type:
Antigen: SCNN1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human