Order Entry
Switzerland
ContactUsLinkComponent
Anti-AHR Rabbit Polyclonal Antibody
Anti-AHR Rabbit Polyclonal Antibody
Catalog # PRSI27-379
Supplier:  ProSci Inc.
undefined
Anti-AHR Rabbit Polyclonal Antibody
Catalog # PRSI27-379
Supplier:  ProSci Inc.

Specifications

  • Antigen name:
    Aryl Hydrocarbon Receptor
  • Antigen symbol:
    AHR
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human,
    Dog,
    Mouse
  • Western blot:
    Yes
  • Form:
    Lyophilized
  • Antigen synonyms:
    BHLHE76|AHR|Ah receptor|AhR|Class E basic helix-loop-helix protein 76|bHLHe76
  • Immunogen:
    A synthetic peptide corresponding a region of human AHR.
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Aryl hydrocarbon receptor (AHR) is a ligand-activated transcription factor involved in the regulation of biological responses to planar aromatic hydrocarbons. AHR has been shown to regulate xenobiotic-metabolizing enzymes such as cytochrome P450. AHR ligands included a variety of aromatic hydrocarbons.

Antibody is purified by protein A chromatography method.

Antibody is lyophilized in PBS buffer with 2% sucrose. Add 100 μL of distilled water. Final antibody concentration is 1 mg/mL.

Aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

Type:
Antigen: AHR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Dog, Human, Mouse