Order Entry
Switzerland
ContactUsLinkComponent
Elemental analysis standards, sulphur standards, CONOSTAN
Elemental analysis standards, sulphur standards, CONOSTAN
Catalog # PROOCON150-400-003
CAS Number:  
Elemental analysis standards, sulphur standards, CONOSTAN
Catalog # PROOCON150-400-003
CAS Number:  

Specifications

  • Matrix:
    Mineral oil
  • Pk:
    100 g
  • Environmentally Preferable:
  • Element:
    Sulphur
  • Symbol:
    S

Specifications

About this item