Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Cimetidine, Reference Standards

Cimetidine, Reference Standards

Supplier:   USP
Cimetidine, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1134062.
  • 1134073.
  • Description:
  • Cimetidine
  • Cimetidine Hydrochloride
  • CAS No.:
  • 51481-61-9
  • 70059-30-2
  • Formula:
  • C₁₀H₁₆N₆S
  • C₁₀H₁₇ClN₆S

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1134062.
    Cimetidine
    Cimetidine
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - SFr. 403.00
    SFr. 403.00
    1 * 200 mg
       SFr. 403.00

    Specifications:
    More
    Specifications
    Cat. No.
    1134062.
    Description
    Cimetidine
    CAS No.
    51481-61-9
    Formula
    C₁₀H₁₆N₆S
  • 1134073.
    Cimetidine Hydrochloride
    Cimetidine Hydrochloride
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - SFr. 409.00
    SFr. 409.00
    1 * 200 mg
       SFr. 409.00

    Specifications:
    More
    Specifications
    Cat. No.
    1134073.
    Description
    Cimetidine Hydrochloride
    CAS No.
    70059-30-2
    Formula
    C₁₀H₁₇ClN₆S