Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Chlorhexidine, Reference Standards

Chlorhexidine, Reference Standards

Supplier:   USP
Chlorhexidine, Reference Standards
Supplier:  USP
Certificates

3 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1111103.
  • 1111012.
  • 1111001.
  • Description:
  • Chlorhexidine Acetate
  • Chlorhexidine System Suitability Mixture
  • Chlorhexidine
  • CAS No.:
  • 56-95-1
  • 18472-51-0
  • 55-56-1
  • Formula:
  • C₂₆H₃₈Cl₂N₁₀O₄
  • C₃₄H₅₄Cl₂N₁₀O₁₄
  • UN No.:
  • 2811
  • 3077
  • 3077

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1111103.
    Chlorhexidine Acetate
    Chlorhexidine Acetate
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - SFr. 462.00
    SFr. 462.00
    1 * 200 mg
       SFr. 462.00

    Specifications:
    More
    Specifications
    Cat. No.
    1111103.
    Description
    Chlorhexidine Acetate
    CAS No.
    56-95-1
    Formula
    C₂₆H₃₈Cl₂N₁₀O₄
    UN No.
    2811
  • 1111012.
    Chlorhexidine System Suitability Mixture
    Chlorhexidine System Suitability Mixture
    USP reference standard
    Vial
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg - SFr. 1’560.00
    SFr. 1’560.00
    1 * 50 mg
       SFr. 1’560.00

    Specifications:
    More
    Specifications
    Cat. No.
    1111012.
    Description
    Chlorhexidine System Suitability Mixture
    CAS No.
    18472-51-0
    Formula
    C₃₄H₅₄Cl₂N₁₀O₁₄
    UN No.
    3077
  • 1111001.
    Chlorhexidine
    Chlorhexidine
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - SFr. 462.00
    SFr. 462.00
    1 * 200 mg
       SFr. 462.00

    Specifications:
    More
    Specifications
    Cat. No.
    1111001.
    Description
    Chlorhexidine
    CAS No.
    55-56-1
    Formula
    UN No.
    3077