Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Chlorthalidone, Reference Standards

Chlorthalidone, Reference Standards

Supplier:   USP
Chlorthalidone, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1119309.
  • 1130006.
  • Description:
  • Chlorthalidone Related Compound A
  • Chlorthalidone
  • Synonym(s):
  • 4'-Chloro-3'-sulfamoyl-2-benzophenone Carboxylic Acid
  • CAS No.:
  • 5270-74-6
  • 77-36-1
  • Formula:
  • C₁₄H₁₁ClN₂O₄S

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1119309.
    Chlorthalidone Related Compound A
    Chlorthalidone Related Compound A
    USP reference standard
    Vial
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg - 1 310.00 SFr.
    1 310.00 SFr.
    1 * 10 mg
       1 310.00 SFr.

    Specifications:
    More
    Specifications
    Cat. No.
    1119309.
    Description
    Chlorthalidone Related Compound A
    Synonym(s)
    4'-Chloro-3'-sulfamoyl-2-benzophenone Carboxylic Acid
    CAS No.
    5270-74-6
    Formula
  • Chlorthalidone
    USP reference standard
    Vial
    200 mg
    Log in for availability
    Log in for availability
    1 * 200 mg
    1 * 200 mg - 440.00 SFr.
    440.00 SFr.
    1 * 200 mg
       440.00 SFr.

    Specifications:
    More
    Specifications
    Cat. No.
    1130006.
    Description
    Chlorthalidone
    Synonym(s)
    CAS No.
    77-36-1
    Formula
    C₁₄H₁₁ClN₂O₄S
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.