Anti-BIRC5 Rabbit Polyclonal Antibody
Catalog # BSBTA00379
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Baculoviral IAP repeat-containing protein 5
- Antigen symbol:BIRC5
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Baculoviral IAP repeat-containing protein 5(BIRC5) detection. Tested with WB, ELISA in Human.
Type: Primary
Antigen: BIRC5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human