Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Amiloride, Reference Standards

Amiloride, Reference Standards

Supplier:   USP
Amiloride, Reference Standards
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard

Documents

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1019701.
  • 1019712.
  • Description:
  • Amiloride Hydrochloride
  • Amiloride Related Compound A
  • Synonym(s):
  • Methyl 3,5-diamino-6-chloropyrazine-2-carboxylate
  • CAS No.:
  • 17440-83-4
  • 1458-01-1
  • Formula:
  • C₆H₈ClN₇O·HCl·₂H₂O
  • C₆H₇ClN₄O₂
  • UN No.:
  • 2811

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1019701.
    Amiloride Hydrochloride
    Amiloride Hydrochloride
    USP reference standard
    Vial
    500 mg
    Log in for availability
    Log in for availability
    1 * 500 mg
    1 * 500 mg - SFr. 453.00
    SFr. 453.00
    1 * 500 mg
       SFr. 453.00

    Specifications:
    More
    Specifications
    Cat. No.
    1019701.
    Description
    Amiloride Hydrochloride
    Synonym(s)
    CAS No.
    17440-83-4
    Formula
    C₆H₈ClN₇O·HCl·₂H₂O
    UN No.
    2811
  • 1019712.
    Amiloride Related Compound A
    Amiloride Related Compound A
    USP reference standard
    Vial
    30 mg
    Log in for availability
    Log in for availability
    1 * 30 mg
    1 * 30 mg - SFr. 1’260.00
    SFr. 1’260.00
    1 * 30 mg
       SFr. 1’260.00

    Specifications:
    More
    Specifications
    Cat. No.
    1019712.
    Description
    Amiloride Related Compound A
    Synonym(s)
    Methyl 3,5-diamino-6-chloropyrazine-2-carboxylate
    CAS No.
    1458-01-1
    Formula
    C₆H₇ClN₄O₂
    UN No.