Order Entry
Switzerland
ContactUsLinkComponent
Magnesium standard solution, 1 mg/L
Magnesium standard solution, 1 mg/L
Catalog # REGEICCS07
Supplier:  REAGECON
CAS Number:  
Magnesium standard solution, 1 mg/L
Catalog # REGEICCS07
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pk:
    100 mL
  • Element:
    Magnesium
  • Symbol:
    Mg

Specifications

About this item

Certifications: Traceable to a NIST.