Oxaliplatin, Reference Standards
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
2 Options Available Below
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- 1481204.
- 1481306.
- Description:
- Oxaliplatin
- Oxaliplatin System Suitability
- Synonym(s):
- [SP-4-2-(1R-trans)]-(1,2-cyclohexanediamine-N,N') dichloridoplatinum(II)
- CAS No.:
- 61825-94-3
- 61848-66-6
- Formula:
- C₈H₁₄N₂O₄Pt
- C₆H₁₄Cl₂N₂Pt
- UN No.:
- 3467
- 3467
Specifications
Product Family Options
Product Information
- StandardsPack typePkAvailabilityPrice
- 1481204.OxaliplatinSpecifications:
More
Specifications
Cat. No.1481204.DescriptionOxaliplatinSynonym(s)CAS No.61825-94-3FormulaC₈H₁₄N₂O₄PtUN No.3467 - 1481306.Oxaliplatin System SuitabilityOxaliplatin System SuitabilityUSP reference standardVial25 mgLog in for availability1 * 25 mg1 410.00 SFr.Specifications:
More
Specifications
Cat. No.1481306.DescriptionOxaliplatin System SuitabilitySynonym(s)[SP-4-2-(1R-trans)]-(1,2-cyclohexanediamine-N,N') dichloridoplatinum(II)CAS No.61848-66-6FormulaC₆H₁₄Cl₂N₂PtUN No.3467



