ContactUsLinkComponent
Quantus® Protect Xtra, Cleanroom Overalls with Adjustable Ankles
Quantus® Protect Xtra, Cleanroom Overalls with Adjustable Ankles
Supplier:  VWR Collection
Certificates
undefined
Quantus® Protect Xtra, Cleanroom Overalls with Adjustable Ankles
Supplier:  VWR Collection
Specifications
  • Catalog No:
  • QNTS081-1PXWH2XLID
  • QNTS081-1PXWH00SID
  • QNTS081-1PXWH00MID
  • QNTS081-1PXWH00LID
  • QNTS081-1PXWH0XLID
  • QNTS081-1PXWH0XSID
  • Model:
  • Protect Xtra
  • Protect Xtra
  • Protect Xtra
  • Protect Xtra
  • Protect Xtra
  • Protect Xtra
  • Garment application:
  • Cleanroom, Laboratory
  • Cleanroom, Laboratory
  • Cleanroom, Laboratory
  • Cleanroom, Laboratory
  • Cleanroom, Laboratory
  • Cleanroom, Laboratory
  • Gender:
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Unisex
  • Sleeve or leg length:
  • Long
  • Long
  • Long
  • Long
  • Long
  • Long
  • Closure type:
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Zip closure
  • Fabric:
  • 100% woven polyester with conductive yarn
  • 100% woven polyester with conductive yarn
  • 100% woven polyester with conductive yarn
  • 100% woven polyester with conductive yarn
  • 100% woven polyester with conductive yarn
  • 100% woven polyester with conductive yarn
  • Weight:
  • 98,3 g/m²
  • 98,3 g/m²
  • 98,3 g/m²
  • 98,3 g/m²
  • 98,3 g/m²
  • 98,3 g/m²