Order Entry
Switzerland
ContactUsLinkComponent
Organic reference standards, trimethoprims, Ph. Eur., RS
Organic reference standards, trimethoprims, Ph. Eur., RS
Supplier:  EDQM
Certificates
Organic reference standards, trimethoprims, Ph. Eur., RS
Supplier:  EDQM

Specifications

  • CAS No.:
    30806-86-1
  • Formula:
    C₁₄H₁₆N₄O₄
  • Description:
    Trimethoprim impurity B

Specifications

Frequently Bought Together