Order Entry
Switzerland
ContactUsLinkComponent
Hand protector
Hand protector
  112-1981
undefined
Hand protector
  112-1981

 

  • Colour:
    Red
  • L×W:
    190×98 mm
  • Seq:
    06
  • Pk:
    1

 

 

This heat resistant hand protector made from silicone rubber protects hands while handling hot labware.

  • Manufactured from strong, tear resistant silicone rubber
  • Withstands temperatures as high as 260 °C and as low as −57 °C
  • Multiple concave tipped studs give a non-slip grip
  • One size