Impurity and API standard, 1,1'-[4,4'bis(methoxy)biphenyl-3,3'-diyl]bis(tricyclo[3.3.1.13,7]decane)
Catalog # PROOMM0681.02
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:50 mg
- Standards:Reference standards
- CAS No.:932033-57-3
- Description:1,1'-[4,4'Bis(methoxy)biphenyl-3,3'-diyl]bis(tricyclo[3.3.1.13,7]decane)
- Environmentally Preferable:
- Formula:C₃₄H₄₂O₂
- Description:1,1'-[4,4'Bis(methoxy)biphenyl-3,3'-diyl]bis(tricyclo[3.3.1.13,7]decane)