Order Entry
Switzerland
ContactUsLinkComponent
Impurity and API standard, 1-benzyl-4-[(5,6-dimethoxy-1H-inden-2-yl)methyl]piperidine
Impurity and API standard, 1-benzyl-4-[(5,6-dimethoxy-1H-inden-2-yl)methyl]piperidine
Catalog # PROOMM0677.16
CAS Number:  
Impurity and API standard, 1-benzyl-4-[(5,6-dimethoxy-1H-inden-2-yl)methyl]piperidine
Catalog # PROOMM0677.16
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Standards:
    Reference standards
  • CAS No.:
    120013-45-8
  • Description:
    1-Benzyl-4-[(5,6-dimethoxy-1H-inden-2-yl)methyl]piperidine
  • Formula:
    C₂₄H₂₉NO₂
  • Description:
    1-Benzyl-4-[(5,6-dimethoxy-1H-inden-2-yl)methyl]piperidine

Specifications

About this item