Order Entry
Switzerland
ContactUsLinkComponent
Impurity and API standard, 1-benzyl-4-[(5,6-dimethoxy-2,3-dihydro-1H-inden-2-yl)methyl]piperidine
Impurity and API standard, 1-benzyl-4-[(5,6-dimethoxy-2,3-dihydro-1H-inden-2-yl)methyl]piperidine
  PROOMM0677.15
 :  
Impurity and API standard, 1-benzyl-4-[(5,6-dimethoxy-2,3-dihydro-1H-inden-2-yl)methyl]piperidine
  PROOMM0677.15
 :  

 

  • Pk:
    50 mg
  • Standards:
    Reference standards
  • CAS No.:
    844694-84-4
  • Description:
    1-Benzyl-4-[(5,6-dimethoxy-2,3-dihydro-1H-inden-2-yl)methyl]piperidine
  • Formula:
    C₂₄H₃₁NO₂
  • Description:
    1-Benzyl-4-[(5,6-dimethoxy-2,3-dihydro-1H-inden-2-yl)methyl]piperidine